missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for HU6 Mini Horizontal Gel Electrophoresis Unit
For use with HU6 Mini Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™ HU6-C1.5-12MC
This item is not returnable.
View return policy
Description
- Color coded
- Height adjustable
- Offer complete control over loading volume
- Well depth to maximum 32-sample throughput
Specifications
HU6 Mini Horizontal Gel Electrophoresis Unit | |
12 | |
17 μL |
Comb | |
1.5 mm | |
2.5 mm |