missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Description
- For tightly resolved bands
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Sub-Gel Midi Horizontal Gel Electrophoresis Unit | |
25 | |
0.75 mm |
Comb | |
1 Each | |
7 μL |