Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£19.05 - £29.60
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11803313
|
Fisherbrand™
MS10-10MC-1 |
10 | 27 μL |
£29.50
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
11833313
|
Fisherbrand™
MS10-1-1 |
1 Prep, 1 Marker | 360 μL |
£19.05
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
11873313
|
Fisherbrand™
MS10-12-1 |
12 | 23 μL |
£29.50
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
11813323
|
Fisherbrand™
MS10-16-1 |
16 | 16 μL |
£29.50
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
11863323
|
Fisherbrand™
MS10-20-1 |
20 | 14 μL |
£19.55
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
11893323
|
Fisherbrand™
MS10-2-1 |
2 Prep, 2 Marker | 158 μL |
£19.05
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
11833333
|
Fisherbrand™
MS10-25-1 |
25 | 10 μL |
£19.55
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
11873333
|
Fisherbrand™
MS10-4-1 |
4 Prep, 2 Marker | 77 μL |
£19.05
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
11813343
|
Fisherbrand™
MS10-8-1 |
8 | 41 μL |
£29.60
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
Description
- Supplied as standard
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Comb | |
1 Each | |
1 mm |