missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 1.5mm thick
For use with HU13 Midi Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™ HU13-C1.5-12MC
This item is not returnable.
View return policy
Description
- Color coded
- Height adjustable
- Offer complete control over loading volume
- Well depth to maximum 112-sample throughput
Specifications
HU13 Midi Horizontal Gel Electrophoresis Unit | |
12 | |
50 μL |
Comb | |
1.5 mm | |
8 mm |