Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 1.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£27.55 - £42.95
Specifications
For Use With (Equipment) | MultiSUB Maxi Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Thickness (Metric) | 1.5 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11863423
|
Fisherbrand™
|
10 | 108 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11893423
|
Fisherbrand™
|
1 Prep, 1 Marker | 1013 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11833433
|
Fisherbrand™
|
16 | 61 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11883433
|
Fisherbrand™
|
20 | 41 μL |
£42.95
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
11813443
|
Fisherbrand™
|
2 Prep, 2 Marker | 473 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11853443
|
Fisherbrand™
|
25 | 32 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11833453
|
Fisherbrand™
|
36 | 22 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11883453
|
Fisherbrand™
|
40 | 17 μL |
£27.55
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
11813463
|
Fisherbrand™
|
4 Prep, 2 Marker | 230 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11853463
|
Fisherbrand™
|
50 | 16 μL |
£27.55
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11893443
|
Fisherbrand™
|
30 | 26 μL |
£28.45
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||||
Description
- Recommended for high resolution gels and publication quality data
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Maxi Horizontal Gel Electrophoresis Unit | |
Comb | |
1 Each | |
1.5 mm |