Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£28.30 - £44.50
Specifications
For Use With (Equipment) | MultiSUB Maxi Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Thickness (Metric) | 2 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11873423
|
Fisherbrand™
|
10 | 144 μL |
£29.05
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||||
11803433
|
Fisherbrand™
|
1 Prep, 1 Marker | 1350 μL |
£30.50
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
11843433
|
Fisherbrand™
|
16 | 81 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11893433
|
Fisherbrand™
|
20 | 54 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11823443
|
Fisherbrand™
|
2 Prep, 2 Marker | 630 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11863443
|
Fisherbrand™
|
25 | 42 μL |
£44.50
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
11803453
|
Fisherbrand™
|
30 | 34 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11843453
|
Fisherbrand™
|
36 | 29 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11893453
|
Fisherbrand™
|
40 | 23 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11823463
|
Fisherbrand™
|
4 Prep, 2 Marker | 306 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
11863463
|
Fisherbrand™
|
50 | 21 μL |
£28.30
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
||||
Description
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Maxi Horizontal Gel Electrophoresis Unit | |
Comb | |
1 Each | |
2 mm |