Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£21.90 - £34.50
Specifications
For Use With (Equipment) | MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Thickness (Metric) | 1 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11843363
|
Fisherbrand™
|
10 | 45 μL |
£34.35
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
11883363
|
Fisherbrand™
|
10 | 29 μL |
£21.90
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11813373
|
Fisherbrand™
|
1 Prep, 1 Marker | 495 μL |
£21.90
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
11853373
|
Fisherbrand™
|
12 | 41 μL |
£24.55
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||||
11893373
|
Fisherbrand™
|
14 | 29 μL |
£21.90
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||||
11833383
|
Fisherbrand™
|
16 | 27 μL |
£27.55
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11873383
|
Fisherbrand™
|
18 | 11 μL |
£21.90
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11823393
|
Fisherbrand™
|
20 | 21 μL |
£34.45
Each |
In Stock
Log in to see stock. |
||||
11853393
|
Fisherbrand™
|
2 Prep, 2 Marker | 225 μL |
£21.90
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
11833403
|
Fisherbrand™
|
30 | 13 μL |
£34.50
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11873403
|
Fisherbrand™
|
35 | 10 μL |
£34.45
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11813413
|
Fisherbrand™
|
4 Prep, 2 Marker | 122 μL |
£21.90
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
Description
- Supplied as standard
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit | |
Comb | |
1 mm |