Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£23.15 - £36.50
Specifications
For Use With (Equipment) | MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Thickness (Metric) | 2 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11863363
|
Fisherbrand™
|
10 | 90 μL |
£23.15
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11803373
|
Fisherbrand™
|
10 | 59 μL |
£23.15
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11833373
|
Fisherbrand™
|
1 Prep, 1 Marker | 990 μL |
£23.15
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11873373
|
Fisherbrand™
|
12 | 81 μL |
£23.15
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
11813383
|
Fisherbrand™
|
14 | 59 μL |
£23.15
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
11853383
|
Fisherbrand™
|
16 | 54 μL |
£36.50
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
11893383
|
Fisherbrand™
|
18 | 23 μL |
£23.15
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11843393
|
Fisherbrand™
|
20 | 43 μL |
£36.40
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
11873393
|
Fisherbrand™
|
2 Prep, 2 Marker | 450 μL |
£23.15
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
11813403
|
Fisherbrand™
|
28 | 23 μL |
£23.15
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11853403
|
Fisherbrand™
|
30 | 25 μL |
£36.50
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
11893403
|
Fisherbrand™
|
35 | 20 μL |
£23.15
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11833413
|
Fisherbrand™
|
4 Prep, 2 Marker | 243 μL |
£23.15
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
Description
- Recommended for high resolution gels and publication quality data
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit | |
Comb | |
1 Each | |
2 mm |