missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Upright Freezers, 100 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
£150.00 - £184.00
Specifications
Height (Metric) | 100 mm (Box) |
---|---|
For Use With (Equipment) | Upright Freezers |
Material | Stainless Steel |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12435830
|
Tenak™
TE24292 |
Boxes (4) |
£150.00
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15200784
|
Tenak™
TE24302 |
Boxes (8) |
£157.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||||
15220784
|
Tenak™
TE24304 |
Boxes (12) |
£166.00
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15260774
|
Tenak™
TE24294 |
Boxes (6) |
£150.00
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15260784
|
Tenak™
TE24312 |
Boxes (15) |
£184.00
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
15280774
|
Tenak™
TE24296 |
Boxes (9) |
£157.00
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
Specifications
100 mm (Box) | |
Upright Freezers | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title