Learn More
Tenak™ Comfort Rack for Upright Freezers, 50 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand:  Tenak™ TE24228
Additional Details : Weight : 4.10000kg
 View more versions of this product
Product Code. 15270734
  • £158.00 / Each
Estimated Shipment: 11-11-2024
to see stock.
Add to basket
Description
Description
  • In the comfort rack, the cryoboxes are placed in drawers
  • Increases the flexibility and reduces the operating weight compared to the rack types
  • Measure the inside height and depth of you freezer when choosing the right rack
  • Racks can be used with all freezer models
  • Constructed of stainless steel
Specifications
Specifications
50 mm (Box)
Boxes (21)
Stainless Steel
Upright Freezers
7 x 3