missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Upright Freezers, 75 mm Box Height, with Boxes
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand: Tenak™ TE24274B
This item is not returnable.
View return policy
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
- Include standard white cardboard boxes with 9 x 9 cell dividers
Specifications
75 mm (Box) | |
Upright Freezers | |
3 x 4 |
562 x 140 x 246 mm | |
Boxes (12) | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tenak™ Comfort Rack for Upright Freezers, 75 mm Box Height, with Boxes > Holds: 12 Boxes (3 x 4)
Spot an opportunity for improvement?Share a Content Correction