missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Compact UV Lamp
Brand: UVP 95-0018-03
Code : 03
Additional Details : Weight : 0.80000kg
Specifications
Compact UV Lamp | |
4 W | |
198 x 71 mm | |
50/60 Hz | |
Plastic | |
71 mm |
230 V | |
365 nm | |
230 V, 50/60 Hz | |
198 mm | |
UVL-21 |