Learn More
3M™ Comply™ Bowie and Dick Autoclave Test Pack
The Bowie and Dick test is carried out daily in an empty chamber and is designed to show the adequacy of steam penetration.
Brand:  3M™ 7000034520
Product Code. 16854273
  • £237.00 / 20 tests
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easy to detect failures according to ISO 11140 Part 1 and Part 3
  • Test pack will give you peace of mind and prove your sterilizer is working adequately
  • Saves time - obvious colour change allows easy interpretation, simple and easy to use
  • Accurate, consistent, reliable and reproducible
  • Reduced storage space - compact and disposable
  • Cost saving - paper can be re-used for note-taking
  • Irreversible, for safe storage and traceability
  • One pack of 20 sheets will last you a month - saving time on placing orders
Specifications
Specifications
EN867-4 and ISO 11140 standards
Autoclave
Unique colour chemistry, disposable
20 Pk.
Steam
For daily autoclave tests
Autoclave Test Pack
Paper
Yes