missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Comply™ Chemical Indicator Strips
Brand: 3M™ 7000002743
This item is not returnable.
View return policy
Specifications
Blue to Pink | |
10 cm | |
Hydrogen Peroxide | |
10 cm |
10 x 10 cm | |
Chemical Indicator Strips | |
1000 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Comply™ Chemical Indicator Strips > Dimensions (L x W): 10 x 10cm
Spot an opportunity for improvement?Share a Content Correction