Learn More
3M™ Comply Disposable Bowie & Autoclave Test Pack
Fast, efficient and easy to use. Paper can be re-used for note-taking
Brand:  3M™ 7000066392
Product Code. 12348199
  • £440.00 / Pack of 20
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
With unique color chemistry, the 3M™ Comply™ disposable Bowie and Dick type autoclave test pack conforms to BS EN 867-4 and carries the BSI KiteMark™.
  • Fast, efficient and disposable
  • Simple and easy to read
  • Stronger colors and higher definition
  • Accurate, consistent, reliable and reproducible
  • Easy record storage
  • Saves time
  • Paper can be re-used for note-taking For single use in porous load sterilisers for daily Bowie and Dick tests.
Specifications
Specifications
Autoclavable
BS EN 867-4
Disposable Bowie and Autoclave Test
20 Pk.