missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Concanavalin A (Con A) Conjugates
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Brand: Invitrogen™ C825
151.24 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).Specifications
Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
Canavlia ensiformis | |
α-man, α-glc | |
Native | |
Store in freezer (-5 to -30°C) and protect from light. | |
Classic Dyes | |
Natural | |
Lyophilized | |
Imaging | |
Texas Red™ | |
Lectin |
10 mg | |
Texas Red | |
C. ensiformis | |
None | |
595/615 nm | |
Room Temperature | |
Lectins | |
Concanavalin A (Con A) | |
Red | |
Imaging Quality |