missing translation for 'onlineSavingsMsg'
Learn More

Genevac Concentrator EZ-2 Plus HCl 2 place swing rotor & 2 swings 597mm x 645mm x 700mm (w x d x h)

Brand:  Genevac EZ3P-23050-HP0

Concentrator EZ-2 Plus HCl 2 place swing rotor & 2 swings 597mm x 645mm x 700mm (w x d x h)

Code : 50

Additional Details : Weight : 88.00000kg

Product Code. 12887643

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers