missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Concentrator Plus
Eppendorf Concentrator plus was designed to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench.
Brand: Eppendorf™ 5305000762
Beschreibung
Spezifikation
Without rotor | |
230 V | |
Concentrator Plus with GB Plug | |
350 W |
Concentrators | |
Integrated diaphragm vacuum pump | |
230 V, 50/60 Hz (GB) | |
58 x 33 x 29 cm (12.6 x 20.9 x 11.7 in.) |