Learn More
Zeiss™ Condenser
Use this condenser for a variety of applications
Brand:  Zeiss™ 424228-9030-000
 View more versions of this product
Product Code. 15254116
  • £1220.00 / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Condenser, achromatic-aplanatic 0.9 H with front lens which can be switched on the right and left
  • Condenser, achromatic-aplanatic 0.9 H D Ph DIC with front lens switchable on the left and right and 5-position modulator disk
  • Condenser 0.9/ 1.25 H, D, Ph1, Ph2, Ph3 for Axio Lab.A1 with low-power system from 2.5x
  • LD condenser 0.4 for slider; For H, Ph, PlasDIC with 10 x 46mm slider mount
Specifications
Specifications
LD Condenser 0.4 for Slider, For H, Ph, PlasDIC
53 mm
10 x 46mm slider mount
0.4 mm
Rotatable, with click stops 0°, 90°, 180°, 270°
Objectives 5x
1 Ea.