missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Condenser

Use this condenser for a variety of applications

£60.00 - £1220.00

Specifications

Quantity 1 Ea.
View More Specs

Products 3
Product Code Brand Type Focus Distance For Use With (Equipment) Includes Price Quantity & Availability  
Product Code Brand Type Focus Distance For Use With (Equipment) Includes Price Quantity & Availability  
15313964
View Documents Promotion Details
Zeiss™
42422790000000
Condenser 0.9/ 1.25 H, D, Ph1, Ph2, Ph3 0.8 mm Axio Lab.A1 With low-power system from 2.5x This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
13488928
View Documents Promotion Details
Zeiss™
424228-9060-000
LD Condenser 0.4 for Slider, For H, Ph, PlasDIC 53 mm Objectives 5x 10 x 46mm slider mount
£60.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
 
15254116
View Documents Promotion Details
Zeiss™
424228-9030-000
LD Condenser 0.4 for Slider, For H, Ph, PlasDIC 53 mm Objectives 5x 10 x 46mm slider mount
£1220.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
 
Description

Description

  • Condenser, achromatic-aplanatic 0.9 H with front lens which can be switched on the right and left
  • Condenser, achromatic-aplanatic 0.9 H D Ph DIC with front lens switchable on the left and right and 5-position modulator disk
  • Condenser 0.9/ 1.25 H, D, Ph1, Ph2, Ph3 for Axio Lab.A1 with low-power system from 2.5x
  • LD condenser 0.4 for slider; For H, Ph, PlasDIC with 10 x 46mm slider mount
Specifications

Specifications

1 Ea.
Videos
SDS
Documents

Documents

Product Certifications