Learn More
Lenz Laborglasintrumente Condensers Acc. to Allihn
Product Code. p-7222460
Click to view available options
Standard Taper Joint No:
NS 19/26
NS 24/29
NS 29/32
No. of Bulbs:
5
6
8
Unit Size:
Each
Product Code. 11856204
Brand: Lenz Laborglasintrumente 05223204
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Allihn condensers acc. to DIN 12581
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Includes Glass olives
Length (Metric) 160 mm
No. of Bulbs 6
Standard Taper Joint No NS 29/32
Material Borosilicate Glass (3.3)