Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn, for Extractors, with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05226005
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10244101
  • £104.00 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Allihn condensers for condensing vapours
  • With NS cone
  • Water connection: olives
  • Made of DURAN™ tubing
Specifications
Specifications
NS 34/35
Duran Glass