Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig, with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£40.00 - £65.00
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
11526794
|
Lenz Laborglasintrumente
05210311 |
NS 14/23 |
£40.00
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
11536794
|
Lenz Laborglasintrumente
05210411 |
NS 13/23 |
£65.00
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
Description
- Liebig condensers acc. to DIN 12576 and in addition to DIN
- For condensing vapours
- With NS socket and NS cone
- Water connection: GL 14 threads (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
GL 14 Threads |