Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05210403
Additional Details : Weight : 0.25000kg
Description
- Condensers acc. to Liebig
- With olives Liebig condenser
- Acc. to DIN 12576
- For condensing vapours
- With NS socket and NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
NS 24/29 | |
Borosilicate Glass (3.3) | |
400 mm |