Learn More
Conductivity Calibration Standards, YSI™
Click to view available options
Quantity:
1 qt.
8 x 1 pt.
Packaging:
Glass Bottle
Plastic Bottle
Conductivity:
1,000μS/cm (Fresh water)
10,000μS/cm
100,000μS/cm
50,000μS/cm (sea water)
Description
- Conductivity solutions are inspected with reference to primary standard solutions according to OIML recommendation 56
- Bottles include a table of corrections at temperatures between 20 and 30°C
- Available as: 1 Quart or in a box of 8 individual pints
Specifications
Packaging | Plastic Bottle |
Quantity | 8 x 1 pt. |
Type | Conductivity Calibration Standard |
Conductivity | 50,000μS/cm (sea water) |