Learn More
Cytiva Conductivity Flow Cell
Brand:  Cytiva 18-1111-05
Code : GX
Additional Details : Weight : 0.16800kg
 View more versions of this product
Product Code. 11330232
  • £1331.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Specifications
UNF 10-32 2 B finger tights
Titanium and Chlorotrifluoroethylene
Conductivity Flow Cell
1 to 14
4.23 mm
Monitor pH/C-900
50 bar
0.041 mL
1 Pack