missing translation for 'onlineSavingsMsg'
Learn More

Consort™ Multi-parameter Analyzer Kits

Includes all the components needed for electrochemical measurement. Consort™ Multi-parameter Analyzer Kits include a high-quality, portable meter as well as the necessary electrodes; buffers, solutions and standards; and adapters and accessories required for field and laboratory use.

£987.47

Specifications

Accuracy (Conductivity) 1%
Accuracy (Temperature) 0.5°C
Description pH/Conductivity/DO kit
Dimensions (L x W x H) 13 x 18 x 10 cm
Inputs BNC (pH, mV, conductivity, DO), Banana for Pt1000 (temperature)
View More Specs

Products 1
Product Code Brand Model Description Includes Price Quantity & Availability  
Product Code Brand Model Description Includes Price Quantity & Availability  
11965458
View Documents
Consort™
C5020T
C5020T pH/Conductivity/DO kit C5020 Meter, pH/ATC electrode SP10T, conductivity/ATC electrode SK10T, DO electrode SZ10T, pH buffers, electrolyte, conductivity standard, mains adapter This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

C5020T: pH/EC/DO Meter Kit

The C5020 is a complete and accurate portable meter that can measure all common electrochemical parameters such as pH, ORP, conductivity, and dissolved oxygen, plus salinity and TDS. It is an all-round instrument with a large conductivity range and measurement storing capabilities. The large graphical display makes it easy to read the measurement, and the stability indicator/algorithm ensures steady, accurate readings.

  • C5020 Multi-Parameter Analyzer
  • Electrodes: pH/ATC electrode SP10T, Conductivity/ATC electrode SK10T, Dissolved oxygen electrode SZ10T
  • 2 x 50mL pH buffers (pH 4 and 7)
  • 50mL Electrolyte solution (3M KCl)
  • 50mL conductivity standard (0.01M KCl)
  • Mains adaptor (100–240 VAC, EU/US)

C6010K: EC Meter Kit

The C6010 is a single channel multi-parameter analyzer suitable as a bench-top or portable device. A corrosion-resistant gold-plated BNC connector ensures a long life and accepts different electrodes. An easy-to-use menu and built-in help system enables users to configure and utilize the instrument successfully. Besides the pre-programmed pH buffers and EC standards, other tables can be added, including the complete temperature table.

  • C6010 Multi-Parameter Analyzer
  • Glass conductivity/ATC electrode SK20T
  • 50mL conductivity standard (0.01M KCl)
  • Carrying case
  • Mains adaptor (100–240 VAC, EU/US)
Specifications

Specifications

1%
pH/Conductivity/DO kit
BNC (pH, mV, conductivity, DO), Banana for Pt1000 (temperature)
C5020T
0 to 1000 mS/cm
0.01 pH
0°C to 100°C
0.5°C
13 x 18 x 10 cm
C5020 Meter, pH/ATC electrode SP10T, conductivity/ATC electrode SK10T, DO electrode SZ10T, pH buffers, electrolyte, conductivity standard, mains adapter
pH/ATC electrode SP10T, conductivity/ATC electrode SK10T, dissolved oxygen electrode SZ10T
0.01 μS/cm
0.1 mg/L
Videos
SDS
Documents

Documents

Product Certifications