missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Resistivity Standard Solution, 133μS/cm, Reagecon™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
500 mL
Specifications
Quantity | 500 mL |
Color | Colorless |
Odor | Characteristic |
Conductivity | 133μS/cm |
Solubility | Completely miscible |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'