missing translation for 'onlineSavingsMsg'
Learn More
Learn More
84 μS/cm Conductivity Standard Solutions, Hanna Instruments™
Click to view available options
Quantity:
500 mL
Description
- Premium quality 84 μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution
- Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
- Air tight bottle with tamper-proof seal to ensure quality
- Premium quality 84 μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material)
- Deionized water for analytical use in accordance with ISO 3696/BS3978
- Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
Specifications
Specifications
Quantity | 500 mL |
Color | Colorless |
Odor | Odorless |
pH | 7 |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'