missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Solution, 50μS.cm-1, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£153.00 - £238.00
Specifications
Conductivity | 50μS.cm-1 |
---|---|
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12954485
|
Fisher Chemical
CTE 004 500 |
500 mL |
£153.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12944485
|
Fisher Chemical
CTE 004 1L |
1000 mL |
£238.00
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
50μS.cm-1 | |
Liquid |