Learn More
Hanna Instruments™ Conductivity Standard Solution
Brand:  Hanna Instruments™ HI-7034
Product Code. 12796955
Description
Description
A choice of ready mixed calibration standards traceable to NIST. Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air. Supplied in boxes of 25.
Specifications
Specifications
∼100°C
Colorless
For laboratory and industrial use
∼7
5 Years
460 mL
Calibration Standard Solutions
80,000 μS/cm
460 mL
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ Conductivity Standard Solution > mV Range: 80000uS/cm; Capacity: 460mL