missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard Solutions, Hanna Instruments™

A premium quality conductivity standard that is prepared against a NIST traceable potassium chloride solution.

Brand:  Hanna Instruments HI70031P

 View more versions of this product

Product Code. 10441141

  • £149.00 / Pack of 25
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Premium quality 1,413 μS/cm conductivity solution that is NIST traceable and contained in portable single-use sachets
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • Convenient packaging for one time use
  • Standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
  • Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
  • A choice of ready mixed calibration standards traceable to NIST
  • Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air
 
Specifications

Specifications

20 mL
Calibration Standard
∼100°C
Laboratory and Industrial Use
Colorless
∼7
1413 μS/cm
Soluble in water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.