missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard Solutions, Hanna Instruments™
A premium quality conductivity standard that is prepared against a NIST traceable potassium chloride solution.
Marke: Hanna Instruments HI7031+COA
Code : 04
Weitere Details : Weight : 0.48960kg
Beschreibung
- A choice of ready mixed calibration standards traceable to NIST
- Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air.
- A premium quality 1,413 μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution
- Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
- Air tight bottle with tamper-proof seal to ensure quality
- Standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
- A choice of ready mixed calibration standards traceable to NIST
- Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air
- Supplied in boxes of 25
Spezifikation
500 mL | |
Calibration Standard | |
∼100°C | |
Laboratory and Industrial Use |
Colorless | |
∼7 | |
1413 μS/cm | |
Soluble in water |