missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard Solutions, Hanna Instruments™
Click to view available options
Quantity:
20 mL
450 mL
500 mL
Conductivity:
12,880 μS/cm
1413 μS/cm
84 μS/cm
Unit Size:
450mL
Each
Pack of 25
Description
- Premium quality 84 μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution
- Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
- Air tight bottle with tamper-proof seal to ensure quality
- 84 μS/cm conductivity solution standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material)
- Deionized water for analytical use in accordance with ISO 3696/BS3978
- Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
- A choice of ready mixed calibration standards traceable to NIST
- Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air
- Supplied in boxes of 25
Specifications
Specifications
| Conductivity | 84 μS/cm |
| Color | Colorless |
| pH | ∼7 |
| Boiling Point | ∼100°C |
| Solubility | Soluble in water |
| Type | Calibration Standard |
| Industry Type | Laboratory Use, Industrial Use |
| Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction