missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard Solutions, Hanna Instruments™

A premium quality conductivity standard that is prepared against a NIST traceable potassium chloride solution.

Brand:  Hanna Instruments HI7031

 View more versions of this product

Product Code. 12776955

  • £116.85 / 450mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A choice of ready mixed calibration standards traceable to NIST
  • Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air.
  • A premium quality 1,413 μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • Air tight bottle with tamper-proof seal to ensure quality
  • Standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
Specifications

Specifications

450 mL
Laboratory and Industrial Use
∼7
Soluble in water
1413 μS/cm
Colorless
∼100°C
Calibration Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Conductivity Standard Solutions, Hanna Instruments™ > mV Range: 1413uS/cm; Capacity: 450mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.