missing translation for 'onlineSavingsMsg'
Learn More
Basic Line Conductivity Standard, 84μS/cm, Hamilton™ Product Code.: 10595673

Basic Line Conductivity Standard, 84μS/cm, Hamilton™

Product Code. 10595673
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 10595673

Brand: Hamilton 238984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 84μS/cm comes with a certificate with calibration document form DFM.

  • Remains stable for up to 3 years
  • Expiration date shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes

Specifications

Packaging 500mL calpack bottle
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Basic Line Conductivity Standard, 84μS/cm, Hamilton™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.