missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, NIST Traceable, 1.30μS/cm, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
250 mL
Description
This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.
- Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
- Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
- Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
- High accuracy (±0.05μS/cm at 25°C)—Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)
Specifications
Specifications
Quantity | 250 mL |
Certificate of Analysis | Yes |
For Use With (Equipment) | Conductivity Meter |
Solvent or Matrix | Aqueous |
Conductivity | 1.30μS/cm (±0.05 at 25°C) |
Physical Form | Liquid |
Documents
Scientific Resources
Brochures
Water analysis
File size: 7.3 MB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
CSKC1325B2 | Certificate of Analysis | 18/02/2025 | CSKC13 |
CSKC1325A2 | Certificate of Analysis | 13/02/2025 | CSKC13 |
CSKC1325A1 | Certificate of Analysis | 28/01/2025 | CSKC13 |
CSKC1324M2 | Certificate of Analysis | 28/01/2025 | CSKC13 |
CSKC1324L3 | Certificate of Analysis | 14/12/2024 | CSKC13 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Conductivity Standard, NIST Traceable, 1.30μS/cm, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction