missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, NIST Traceable, 10,000μS/cm, Reagecon™
BIOPHARMA

Product Code. 11536723
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Accredited to ISO 17025. Reagecon™ Conductivity Standard, NIST Traceable, 10,000μS/cm is an aqueous-based, non-hazardous standard with a high degree of accuracy, low temperature variation and guaranteed stability through the expiration date, even after opening the bottle.

This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.

  • Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
  • Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
  • Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
  • High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)

Specifications

Quantity 500 mL
Certificate of Analysis Yes
For Use With (Equipment) Conductivity Meter
Solvent or Matrix Aqueous
Conductivity 10,000μS/cm (±1% at 25°C)
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'