missing translation for 'onlineSavingsMsg'
Learn More
Conductivity Standard, NIST Traceable, 5μS/cm, Reagecon™ Product Code.: 10460538

Conductivity Standard, NIST Traceable, 5μS/cm, Reagecon™
BIOPHARMA

Product Code. 10460538 Shop All Reagecon Products
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 10460538

Brand: Reagecon CSKC5

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Accredited to ISO 17025. Reagecon™ Conductivity Standard, NIST Traceable, 5μS/cm is an aqueous-based, non-hazardous low-level conductivity standard with a high degree of accuracy, low temperature variation and guaranteed stability through expiration, even after opening the bottle.

This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.

  • Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
  • Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
  • Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
  • High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)

Specifications

Quantity 500 mL
Certificate of Analysis Yes
Conductivity 5μS/cm (±1% at 25°C)
Physical Form Liquid
For Use With (Equipment) Conductivity Meter
Solvent or Matrix Aqueous

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCS525C3 Certificate TypeCertificate of Analysis Date23/04/2025 Catalog NumberCSKC5
Lot NumberCS525C1 Certificate TypeCertificate of Analysis Date01/04/2025 Catalog NumberCSKC5
Lot NumberCS525B3 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberCSKC5
Lot NumberCS525B2 Certificate TypeCertificate of Analysis Date19/02/2025 Catalog NumberCSKC5
Lot NumberCS525A4 Certificate TypeCertificate of Analysis Date19/02/2025 Catalog NumberCSKC5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Conductivity Standard, NIST Traceable, 5μS/cm, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.