missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Test Solution Potassium Chloride, SI Analytics™
Click to view available options
Quantity:
20 mL
250 mL
500 mL
Packaging:
3 x 6 Ampules
Polyethylene Bottle
Concentration:
0.01/0.1/1mol/L (1.41/12.9/112mS/cm)
0.01mol/L (1.41 mS/cm)
0.1mol/L (12.9mS/cm)
5mS/cm
Description
- Mixture consisting of water and potassium chloride
- Store at room temperature
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
- Product does not present an explosion hazard
- Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
- Small amounts may be diluted with plenty of water and washed away
Specifications
Concentration | 5mS/cm |
Packaging | Polyethylene Bottle |
Quantity | 500 mL |
Color | Colorless |
Odor | Odorless |
Density | 1g/cm3 |
pH | 7 |
Melting Point | 0°C |
Boiling Point | 100°C |
Solubility | Miscible with water |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'