missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Cone™ Weather-resistant Blank Cones
Made of high impact, weather-resistant materials, the Cone™ Warning System is an ideal solution for your indoor or outdoor hazardous warning needs. Cone™ Weather-resistant Blank Cones are four-sided, so your message can't be missed.
£55.55 - £75.80
Specifications
Material | Plastic |
---|---|
Displayed Text | Blank |
Length (English) | 10.5 in. |
Length (Metric) | 266.7 mm |
Packaging Quantity | 1 Ea. |
Product Code | Brand | Color | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Height (Metric) | Price | Quantity & Availability | ||||
11791925
|
Brady™
077198 |
Yellow | 635 mm |
£55.55
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11711935
|
Brady™
077204 |
Orange | 889 mm |
£75.80
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11721935
|
Brady™
077205 |
Yellow | 889 mm |
£75.80
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
Description
- Made of high impact, weather-resistant materials.
- Ideal for your indoor or outdoor hazardous warning needs.
- Cones are four-sided, so your message can't be missed.
Specifications
Plastic | |
10.5 in. | |
1 Ea. |
Blank | |
266.7 mm |