missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Congo Red, Indicator grade
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >360.0°C |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol, slightly soluble in acetone, practically insoluble in ether |
Formula Weight | 696.65 |
Grade | Indicator |
Packaging | Glass bottle |
Quantity | 100 g |
Chemical Name or Material | Congo Red |
Safety and Handling
Product Identifier
- Congo red
- Danger
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- H361d-Suspected of damaging the unborn child.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'