Learn More
JG Finneran Associates™ Conical Inserts
Use these inserts for a variety of applications. JG Finneran™ Conical Inserts feature a polymer spring which acts as a shock absorber and protects against breakage if the needle bottoms out.
Brand:  JG Finneran Associates™ 401PBS-530
Code : 17
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12377009
  • £68.65 / Pack of 100
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • Available preassembled with a polymer bottom spring or patented Top Spring™
  • Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
  • Conical design permits complete sample removal
Specifications
Specifications
Polypropylene
Polymer Bottom Spring
100 Pk.
100 μL
Limited volume, precision formed interior