missing translation for 'onlineSavingsMsg'
Learn More

Coniferyl alcohol, 98%

Product Code. 11411487
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11411487

Brand: Thermo Scientific Alfa Aesar B24949.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 458-35-5 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458-35-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00002922
InChI Key JMFRWRFFLBVWSI-NSCUHMNNSA-N
Synonym coniferyl alcohol, coniferol, coniferylalcohol, 4-3-hydroxyprop-1-en-1-yl-2-methoxyphenol, gamma-hydroxyisoeugenol, trans-coniferyl alcohol, e-coniferyl alcohol, 4-3-hydroxy-1-propenyl-2-methoxyphenol, 3-4-hydroxy-3-methoxyphenyl-2-propen-1-ol, 4-1e-3-hydroxyprop-1-en-1-yl-2-methoxyphenol
PubChem CID 1549095
ChEBI CHEBI:17745
SMILES COC1=CC(\C=C\CO)=CC=C1O

Specifications

Melting Point 75°C to 76°C
Boiling Point 163°C to 165°C (3 mmHg)
Quantity 5 g
Beilstein 2048961
Sensitivity Air and light sensitive
Merck Index 14,2504
IUPAC Name 4-[(E)-3-hydroxyprop-1-enyl]-2-methoxyphenol
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Coniferyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.