missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Interface Converter Connection Cable Set
Click to view available options
Voltage:
120 V
230 V
Description
- Set of connection cables and interface connector RS 422 or RS 232
- For either 10 interfaces or one unit
Specifications
Description | Interface converter - RS 422 cable set and RS 232 / RS 422 interface converter for connection to 10-way plug distributor or for connection to a unit |
For Use With (Equipment) | Multi Management Software APT-COM™ |
Type | Accessory, Connection Cable Set |
Electrical Requirements | 120 V, 60 Hz |
Voltage | 120 V |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'