missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ Connection For Test Tubes

Velp Scientifica™ Connection For Test Tubes For use with UDK Distillation Units.

Brand:  Velp Scientifica™ A00000043

Product Code. 12878343

  • £50.25 / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with UDK Distillation Units
Specifications

Specifications

48 mm and 500 mL Kjeldahl Balloon
1 Each
26 mm
UDK127 Kjeldahl Balloon 26, 48 mm dia.
Connection For Test Tubes
1Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers