Learn More
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
10404332
|
Gilson™
F1825056 |
4 mm |
£200.00
Pack of 5 |
In Stock
Log in to see stock. |
|||||
10113002
|
Gilson™
F1825054 |
3 mm |
£212.00
Pack of 5 |
In Stock
Log in to see stock. |
|||||
Description
- Connector kit accessories for the MINIPULS™ 3 and Evolution peristaltic pumps come in a variety of types: metal sleeve connectors for small bore tubing, connector kits for flanged tubing, or plastic connectors for large bore tubing.
- These kits contain five connectors each.
- The connectors are available in a three sizes to fit a variety of tubing of different inner diameters.
Specifications
5 Set | |
Connection Kit |