Learn More
Mettler Toledo™ Connector Syringe/Filling Tube
Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
Densito and DensitoPro Handheld Density Meters
Unit Size:
Each
Description
- When working with viscous samples, this connector acts as an adapter between the Density2Go handheld meter and the syringe, making your work easier
Specifications
Specifications
| Description | Connector Syringe/Filling Tube, Densito |
| Languages | Chinese, English (GB), English (US), French, German, Italian, Korean, Polish, Portuguese (BR), Russian, Spanish |
| Product Type | Accessory |
| For Use With (Equipment) | Densito and DensitoPro Handheld Density Meters |
| For Use With (Application) | Use with Densito and DensitoPro Handheld Density Meters |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu