Learn More
SCAT Connectors for HPLC
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
Brand:  SCAT 107814
8.86 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10349637
  • £10.95 / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Specifications
Tube Connector
Polypropylene
HPLC/UHPLC Systems
5 to 7 mm
Waste Collector Cap and Tubing