Learn More
Specac™ Constant Thickness Film Maker Kit II
Brand: Specac™
Code : Z3
Additional Details : Weight : 69.15000kg
Description
This kit includes a 15 ton manual hydraulic press GS15011, film maker accessory GS15640 and heated platens GS15515 for the preparation of thin films using temperatures up to 300°C.Specifications
15 ton manual hydraulic press GS15011, film maker accessory GS15640 and heated platens GS15515 for the preparation of thin films using temperatures up to 300°C |